Learn More
5alpha-Pregnan-11beta,17alpha,21-triol-3,20-dione-1,2,4,5-d4, CDN
Specifications
Specifications
| Chemical Name or Material | 5 alpha-Dihydrocortisol-1,2,4 alpha,5 alpha-D4 |
| CAS | 252976-11-7 |
| Molecular Formula | C21 D4 H28 O5 |
| Quantity | 0.01 g |
| Synonym | Pregnane-3,20-dione-1,2,4,5-d4, 11,17,21-trihydroxy-, (4α,5α,11β)- (9 CI), (4α,5α,11β)-11,17,21-Trihydroxypregnane-3,20-dione-1,2,4,5-d4, 11β,17,21-Trihydroxy-5α-pregnane-3,20-dione-1,2,4α,5α-d4, 5α-Dihydrocortisol-d4, 5α-Dihydrocortisol-1,2,4α,5α-d4, 5α-Dihydrocortisol-1,2,4α,5α-D4, 5α-Pregnan-11β,17α,21-triol-3,20-dione-1,2,4α,5-d4 |
| InChI Formula | InChI=1 S/C21H32O5/c1-19-7-5-13(23)9-12(19)3-4-14-15-6-8-21(26,17(25)11-22)20(15,2)10-16(24)18(14)19/h12,14-16,18,22,24,26 H,3-11H2,1-2H3/t12-,14-,15-,16-,18+,19-,20-,21-/m0/s1/i5D,7 D,9 D,12 D/t5?,7?,9-,12-,14-,15-,16-,18+,19-,20-,21- |
| SMILES | [2 H]C1C([2 H])[C@]2(C)[C@H]3 [C@@H](O)C[C@@]4(C)[C@@H](CC[C@]4(O)C(=O)CO)[C@@H]3 CC[C@@]2([2 H])[C@H]([2 H])C1=O |
| IUPAC Name | (4 S,5 S,8 S,9 S,10 S,11 S,13 S,14 S,17 R)-1,2,4,5-tetradeuterio-11,17-dihydroxy-17-(2-hydroxyacetyl)-10,13-dimethyl-1,2,4,6,7,8,9,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-3-one |
| Molecular Weight (g/mol) | 368.5004 |
| Formula Weight | 368.2501 g/mol |
| Show More |