missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5beta-Androstan-3alpha-ol-11,17-dione-9,12,12,16,16-d5, CDN
Click to view available options
Quantity:
0.005 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 3 alpha-Hydroxy-5 beta-androstane-11,17-dione-9,12,12,16,16-D5 |
| Molecular Formula | C19 D5 H23 O3 |
| Quantity | 0.005 g |
| Synonym | (3α,5β)-3-Hydroxyandrostane-11,17-dione-9,12,12,16,16-d5, 3α-Hydroxy-5β-androstane-11,17-dione-9,12,12,16,16-D5, 11-Ketoetiocholanolone-d5, 11-Oxoetiocholanolone-d5, 5β-Androstan-3α-ol-11,17-dione-9,12,12,16,16-d5 |
| InChI Formula | InChI=1 S/C19H28O3/c1-18-8-7-12(20)9-11(18)3-4-13-14-5-6-16(22)19(14,2)10-15(21)17(13)18/h11-14,17,20 H,3-10H2,1-2H3/t11-,12-,13+,14+,17-,18+,19+/m1/s1/i6D2,10D2,17 D |
| SMILES | [2 H]C1([2 H])C[C@H]2 [C@@H]3 CC[C@@H]4 C[C@H](O)CC[C@]4(C)[C@@]3([2 H])C(=O)C([2 H])([2 H])[C@]2(C)C1=O |
| IUPAC Name | (3 R,5 R,8 S,9 S,10 S,13 S,14 S)-9,12,12,16,16-pentadeuterio-3-hydroxy-10,13-dimethyl-1,2,3,4,5,6,7,8,14,15-decahydrocyclopenta[a]phenanthrene-11,17-dione |
| Molecular Weight (g/mol) | 309.4546 |
| Formula Weight | 309.2352 g/mol |
| Percent Purity | 91 atom % D, min 98% Chemical Purity |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction