Learn More
Trajan Scientific and Medical™ 5F-PE-GT-0.47 GC Autosampler Syringes
Click to view available options
Volume (Metric):
5 μL
Unit Size:
Each
Description
- Specifically designed for PerkinElmer instruments
Specifications
Specifications
| Material | Borosilicate Glass (Barrel), PTFE (Plunger) |
| Needle Gauge | 26 G |
| Diameter (Metric) Inner Needle | 0.15 mm |
| Diameter (Metric) Outer | 8 mm |
| Diameter (Metric) Outer Needle | 0.47 mm |
| Product Type | GC Autosampler Syringe |
| Model | 5F-PE-GT-0.47 |
| Volume (Metric) | 5 μL |