missing translation for 'onlineSavingsMsg'
Learn More

Androstanolone, TRC

Product Code. 30338160
Click to view available options
Quantity:
5 g
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 30338160

Brand: Toronto Research Chemicals TRCA6375405G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 5a-Dihydrotestosterone (DHT)
CAS 521-18-6
Purity Grade Notes HPLC
Molecular Formula C19 H30 O2
Quantity 5 g
Synonym 5α-Androstan-3-one, 17β-hydroxy- (8CI), (5α,17β)-17-Hydroxyandrostan-3-one, (+)-Androstan-17β-ol-3-one, 17β-Hydroxy-3-androstanone, 17β-Hydroxy-5α-androstan-3-one, 17β-Hydroxy-5α-androstane-3-one, 4,5α-Dihydrotestosterone, 4-Dihydrotestosterone, 5-DHT, 5α,17β-Hydroxyandrostan-3-one, 5α-Androstan-17β-ol-3-one, 5α-Androstanolone, 5α-Dihydrotestosterone, Anaboleen, Anabolex, Andractim, Androlone, Androstan-17β-ol-3-one, Androstanolone, Cristerona MB, DHT, Dihydrotestosterone, LG 152, NSC 10972, Neodrol, Stanaprol, Stanolone, Testosterone, dihydro-
InChI Formula InChI=1S/C19H30O2/c1-18-9-7-13(20)11-12(18)3-4-14-15-5-6-17(21)19(15,2)10-8-16(14)18/h12,14-17,21H,3-11H2,1-2H3/t12-,14-,15-,16-,17-,18-,19-/m0/s1
SMILES C[C@]12CC[C@H]3[C@@H](CC[C@H]4CC(=O)CC[C@]34C)[C@@H]1CC[C@@H]2O
IUPAC Name (5S,8R,9S,10S,13S,14S,17S)-17-hydroxy-10,13-dimethyl-1,2,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydrocyclopenta[a]phenanthren-3-one
Molecular Weight (g/mol) 290.44
Formula Weight 290.2246
Percent Purity >95
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.