missing translation for 'onlineSavingsMsg'
Learn More

5H-Dibenzo[a,d]cycloheptene, TRC

Product Code. 30009662
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009662 250 mg 250mg
30138082 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009662 Supplier Toronto Research Chemicals Supplier No. TRCD417055250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 256-81-5
Formula Weight 192.0939
InChI Formula InChI=1S/C15H12/c1-3-7-14-11-15-8-4-2-6-13(15)10-9-12(14)5-1/h1-10H,11H2
IUPAC Name 11H-dibenzo[1,2-b:2',1'-e][7]annulene
Molecular Formula C15 H12
Molecular Weight (g/mol) 192.26
SMILES C1c2ccccc2C=Cc3ccccc13
Synonym 5H-Dibenzo[a,d][7]annulene, Cyproheptadine Hydrochloride Imp. A (EP), Dibenzocycloheptene
Chemical Name or Material Dibenzocycloheptene

Specifications

Quantity 250 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.