Learn More
Enzo Life Sciences 5mg:Cerulenin
Brand:  Enzo Life Sciences BML-G237-0005
Encompass_Preferred
5mg:Cerulenin
Product Code. 15124888
  • € 181.50 / 5mg
Description
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Cerulenin
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.