Learn More
Enzo Life Sciences 5MG Clozapine N-oxide
Brand:  Enzo Life Sciences BML-NS105-0005
Encompass_Preferred
5MG Clozapine N-oxide
Product Code. 15145428
  • € 113.74 / 5mg
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Clozapine N-oxide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement