Learn More
Enzo Life Sciences 5mg Ochratoxin A
Brand:  Enzo Life Sciences ALX-630-089-M005
Encompass_Preferred
5mg Ochratoxin A
Product Code. 15190698
  • € 469.48 / 5mg
Videos
Safety and Handling
Safety and Handling
Product Identifier
  • Ochratoxin A
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Carcinogenicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P264-Wash thoroughly after handling.