missing translation for 'onlineSavingsMsg'
Learn More

Uridine Diphosphate Choline Ammonium Salt, Uridine Diphosphate Choline Ammonium Salt, TRC

Product Code. 17949094
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
Unit Size:
1mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17949094 5 mg 5mg
17948224 50 mg 50mg
17948324 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17949094 Supplier Toronto Research Chemicals Supplier No. TRCU8299305MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C14H28N4O12P2
Molecular Weight (g/mol) 506.34
Synonym Uridine 5'-(Trihydrogen Diphosphate) P'-[2-(Trimethylammonio)ethyl] Ester Ammonium Salt;
IUPAC Name ((2R,3S,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-3,4-dihydroxytetrahydrofuran-2-yl)methyl trihydrogen diphosphate
SMILES O[C@@H]1[C@@H]([C@H](O[C@H]1N(C(N2)=O)C=CC2=O)COP(OP(OCC[N+](C)(C)C)([O-])=O)([O-])=O)O.[NH4+]

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H25N3O12P2.H3N/c1-17(2,3)6-7-26-30(22,23)29-31(24,25)27-8-9-11(19)12(20)13(28-9)16-5-4-10(18)15-14(16)21;/h4-5,9,11-13,19-20H,6-8H2,1-3H3,(H2-,15,18,21,22,23,24,25);1H3/t9-,11-,12-,13-;/m1./s1
Formula Weight 506.12
Percent Purity >95%
Chemical Name or Material Uridine Diphosphate Choline Ammonium Salt
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.