missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xanthorrhizol (Technical grade), TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | Xanthorrhizol |
| CAS | 30199-26-9 |
| Molecular Formula | C15 H22 O |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C15H22O/c1-11(2)6-5-7-12(3)14-9-8-13(4)15(16)10-14/h6,8-10,12,16H,5,7H2,1-4H3/t12-/m1/s1 |
| SMILES | C[C@H](CCC=C(C)C)c1ccc(C)c(O)c1 |
| IUPAC Name | 2-methyl-5-[(2R)-6-methylhept-5-en-2-yl]phenol |
| Molecular Weight (g/mol) | 218.33 |
| Formula Weight | 218.1671 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction