missing translation for 'onlineSavingsMsg'
Learn More

5T4 Antibody (39A11G5F2) [DyLight 550] - Chimeric, Novus Biologicals™

Product Code. 30532994 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30532994 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30532994 Supplier Novus Biologicals Supplier No. NBP345123R

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

5T4 Monoclonal antibody specifically detects 5T4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen 5T4
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone 39A11G5F2
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, trophoblast glycoprotein
Host Species Rabbit
Immunogen BALB/cAnNCrl mice or SJL/JorllcoCrl mice were immunized with either full length 5T4 or amino acid sequence 32-355 from 5T4 (Uniprot#: Q13641)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology, Stem Cells
Primary or Secondary Primary
Gene ID (Entrez) 7162
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.