missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5T4 Antibody (Wyeth patent anti-5T4) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28734JF669
This item is not returnable.
View return policy
Description
5T4 Monoclonal antibody specifically detects 5T4 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| 5T4 | |
| Monoclonal | |
| Janelia Fluor 669 | |
| 5T4 oncofetal trophoblast glycoprotein, 5T4 oncotrophoblast glycoprotein, 5T4AG, 5T4-AG, 5T4M6P1,5T4 oncofetal antigen, trophoblast glycoprotein | |
| TPBG | |
| 0.1 mL | |
| Cancer, Cell Biology, Immunology, Stem Cells | |
| 7162 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Wyeth patent anti-5T4 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction