missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 5T4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116062
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total TPBG.
This gene encodes a leucine-rich transmembrane glycoprotein that may be involved in cell adhesion. The encoded protein is an oncofetal antigen that is specific to trophoblast cells. In adults this protein is highly expressed in many tumor cells and is associated with poor clinical outcome in numerous cancers. Alternate splicing in the 5' UTR results in multiple transcript variants that encode the same protein. [provided by RefSeq, Oct 2009].
Specifications
| 5T4 | |
| Polyclonal | |
| Unconjugated | |
| TPBG | |
| 5T4; 5T4 oncofetal antigen; 5T4 oncofetal trophoblast glycoprotein; 5T4 oncotrophoblast glycoprotein; 5T4AG; 5T4-AG; AW495680; M6P1; TPBG; Trophoblast glycoprotein; WAIF1; Wnt-activated inhibitory factor 1 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 21983, 7162 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| Q13641, Q9Z0L0 | |
| TPBG | |
| A synthesized peptide derived from human TPBG(Accession Q13641), corresponding to amino acid residues A364-L414. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction