missing translation for 'onlineSavingsMsg'
Learn More

6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number 11996091
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
918643-56-8
C16H27BN2O3
306.21
MFCD06797989
QWJUJXPYTLOKRL-UHFFFAOYSA-N
n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propan-1-amine, 2-3-n,n-dimethylamino-propoxy pyridine-5-boronic acid, pinacol ester, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yloxy propan-1-amine, dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yloxy-propyl-amine, dimethy 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-22-yloxy propyl amine, dimethyl 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propyl amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl oxy-1-propanamine
17998906
N,N-dimethyl-3-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]oxypropan-1-amine
CN(C)CCCOC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1
This item is not returnable. View return policy
918643-56-8
C16H27BN2O3
306.21
MFCD06797989
QWJUJXPYTLOKRL-UHFFFAOYSA-N
n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propan-1-amine, 2-3-n,n-dimethylamino-propoxy pyridine-5-boronic acid, pinacol ester, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yloxy propan-1-amine, dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yloxy-propyl-amine, dimethy 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-22-yloxy propyl amine, dimethyl 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propyl amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl oxy-1-propanamine
17998906
N,N-dimethyl-3-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]oxypropan-1-amine
CN(C)CCCOC1=NC=C(C=C1)B1OC(C)(C)C(C)(C)O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 34°C to 37°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 306.21
Percent Purity 97%
Chemical Name or Material 6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester
compliance-icons
Product Identifier
  • 6-¢3-(Dimethylamino)propoxy!pyridine-3-boronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'