missing translation for 'onlineSavingsMsg'
Learn More

6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester, 97%, ACROS Organics™

Brand:  Acros Organics 444970010

Additional Details : CAS : 918643-56-8 Weight : 0.05100kg


Quantity 1g

 View more versions of this product

Product Code. 11986091

  • € 252.00 / 1g


Estimated Shipment: 16-08-2021
Add to basket

Description

Description

Specifications

Chemical Identifiers

918643-56-8
306.21
n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propan-1-amine, 2-3-n,n-dimethylamino-propoxy pyridine-5-boronic acid, pinacol ester, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yloxy propan-1-amine, dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yloxy-propyl-amine, dimethy 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-22-yloxy propyl amine, dimethyl 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propyl amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl oxy-1-propanamine
N,N-dimethyl-3-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]oxypropan-1-amine
C16H27BN2O3
QWJUJXPYTLOKRL-UHFFFAOYSA-N
17998906
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCCCN(C)C

Specifications

6-[3-(Dimethylamino)propoxy]pyridine-3-boronic acid pinacol ester
34°C to 37°C
918643-56-8
C16H27BN2O3
QWJUJXPYTLOKRL-UHFFFAOYSA-N
N,N-dimethyl-3-[5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]oxypropan-1-amine
17998906
97%
Glass Bottle
1g
96% min. (GC)
n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propan-1-amine, 2-3-n,n-dimethylamino-propoxy pyridine-5-boronic acid, pinacol ester, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yloxy propan-1-amine, dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridin-2-yloxy-propyl-amine, dimethy 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-22-yloxy propyl amine, dimethyl 3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridin-2-yl oxy propyl amine, n,n-dimethyl-3-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2-pyridinyl oxy-1-propanamine
B1(OC(C(O1)(C)C)(C)C)C2=CN=C(C=C2)OCCCN(C)C
306.21
306.21
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 6-¢3-(Dimethylamino)propoxy!pyridine-3-boronic acid pinacol ester

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS Documents
Certificates