Learn More
6,7-Dimethoxy-2-(piperazin-1-yl)quinazolin-4-amine, Mikromol™
Description
6,7-Dimethoxy-2-(piperazin-1-yl)quinazolin-4-amine
Specifications
Specifications
| Chemical Name or Material | 6,7-Dimethoxy-2-(piperazin-1-yl)quinazolin-4-amine |
| CAS | 60547-97-9 |
| Analyte or Component Names | 6,7-Dimethoxy-2-(piperazin-1-yl)quinazolin-4-amine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H19 N5 O2 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Doxazosin Mesilate Imp. G (EP), Prazosin Hydrochloride Imp. C (EP), Prazosin Imp. C (EP), Terazosin Hydrochloride Dihydrate Imp. C (EP), Terazosin Imp. C (EP), Terazosin USP Related Compound A, Terazosin USP RC A, 6,7-Dimethoxy-2-(piperazin-1-yl)quinazolin-4-amine, 4-Quinazolinamine, 6,7-dimethoxy-2-(1-piperazinyl)-, 6,7-Dimethoxy-2-(1-piperazinyl)-4-quinazolinamine, 2-(1-Piperazinyl)-4-amino-6,7-dimethoxyquinazoline, 2-(4-Piperazinyl)-4-amino-6,7-dimethoxyquinazoline, 2-Piperazino-6,7-dimethoxy-4-quinazolinamine, 4-Amino-2-(1-piperazinyl)-6,7-dimethoxyquinazoline, 4-Amino-6,7-dimethoxy-2-(1-piperazinyl)quinazoline, Terazosin Related Compound A, Doxazosin Mesilate Impurity G, Prazosin Hydrochloride Impurity C, Terazosin Hydrochloride Dihydrate Impurity C, Doxazosin Impurity G, Prazosin Impurity C, Terazosin Impurity C |
| InChI Formula | InChI=1S/C14H19N5O2/c1-20-11-7-9-10(8-12(11)21-2)17-14(18-13(9)15)19-5-3-16-4-6-19/h7-8,16H,3-6H2,1-2H3,(H2,15,17,18) |
| SMILES | COc1cc2nc(nc(N)c2cc1OC)N3CCNCC3 |
| IUPAC Name | 6,7-dimethoxy-2-piperazin-1-ylquinazolin-4-amine |
| Show More |