Learn More
6'-Amino-N-[2-(4-morpholinyl)oxazolo[4,5-b]pyridin-6-yl][2,3'-bipyridine]-6-carboxamide, TRC
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Chemical Name or Material | 6'-Amino-N-[2-(4-morpholinyl)oxazolo[4,5-b]pyridin-6-yl][2,3'-bipyridine]-6-carboxamide |
| CAS | 1801343-74-7 |
| Molecular Formula | C21H19N7O3 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C21H19N7O3/c22-18-5-4-13(11-23-18)15-2-1-3-16(26-15)20(29)25-14-10-17-19(24-12-14)27-21(31-17)28-6-8-30-9-7-28/h1-5,10-12H,6-9H2,(H2,22,23)(H,25,29) |
| SMILES | Nc1ccc(cn1)c2cccc(n2)C(=O)Nc3cnc4nc(oc4c3)N5CCOCC5 |
| IUPAC Name | 6-(6-aminopyridin-3-yl)-N-(2-morpholin-4-yl-[1,3]oxazolo[4,5-b]pyridin-6-yl)pyridine-2-carboxamide |
| Molecular Weight (g/mol) | 417.42 |
| Formula Weight | 417.1549 |