Learn More
6-Aminochrysene, MedChemExpress
Product Code. 16402986
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402986 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16402986 Supplier Medchem Express Supplier No. HY108315
MedChemExpress 6-Aminochrysene (6-Aminochrysene) is an aromatic amine used as a chemotherapeutic agent in the treatment of splenomegaly, myeloid leukemia, and breast cancer.
6-Aminochrysene (6-Aminochrysene) is an aromatic amine used as a chemotherapeutic agent in the treatment of splenomegaly, myeloid leukemia, and breast cancer.
Specifications
Chemical Name or Material 6-Aminochrysene
Physical Form Solid
CAS 2642-98-0
Color Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C18H13N
For Use With (Application) Cancer-programmed cell death
Synonym 6-Chrysenamine
Solubility Information DMSO : 250 mg/mL (1027.54 mM; Need ultrasonic)
SMILES NC1=C2C=CC=CC2=C3C(C4=CC=CC=C4C=C3)=C1
Molecular Weight (g/mol) 243.3
Formula Weight 243.3
Percent Purity 95.0%
Grade Research
Show More Show Less
Research purposes only