Learn More
6-Aminochrysene, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Aminochrysene |
| CAS | 2642-98-0 |
| Molecular Formula | C18 H13 N |
| Quantity | 100 mg |
| Synonym | 6-Chrysenamine (6CI, 7CI, 8CI, 9CI, ACI), 6-Aminochrysene, Chrysenex, Chrysonex, NSC 80186 |
| InChI Formula | InChI=1S/C18H13N/c19-18-11-17-13-6-2-1-5-12(13)9-10-15(17)14-7-3-4-8-16(14)18/h1-11H,19H2 |
| SMILES | Nc1cc2c3ccccc3ccc2c4ccccc14 |
| IUPAC Name | chrysen-6-amine |
| Molecular Weight (g/mol) | 243.3 |
| Formula Weight | 243.1048 |