missing translation for 'onlineSavingsMsg'
Learn More

6-Benzyloxypyridine-3-boronic acid, 97%, Thermo Scientific™

Catalog Number p-4380050
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
929250-35-1
C12H12BNO3
229.04
MFCD06798262
CUZOGAUUSKBDAO-UHFFFAOYSA-N
2-benzyloxypyridine-5-boronic acid, 6-benzyloxypyridine-3-boronic acid, 6-benzyloxy pyridin-3-yl boronic acid, 6-benzyloxy pyridine-3-boronic acid, 6-benzyloxy pyridin-3-ylboronic acid, 2-benzyloxy-pyridine-5-boronic acid, 6-benzyloxy pyridin-3-yl-3-boronic acid, 2-benzyloxypyridin-5-yl boronic acid, 2-benzyloxy pyridine-5-ylboronic acid
17750216
(6-phenylmethoxypyridin-3-yl)boronic acid
OB(O)C1=CN=C(OCC2=CC=CC=C2)C=C1
This item is not returnable. View return policy
929250-35-1
C12H12BNO3
229.04
MFCD06798262
CUZOGAUUSKBDAO-UHFFFAOYSA-N
2-benzyloxypyridine-5-boronic acid, 6-benzyloxypyridine-3-boronic acid, 6-benzyloxy pyridin-3-yl boronic acid, 6-benzyloxy pyridine-3-boronic acid, 6-benzyloxy pyridin-3-ylboronic acid, 2-benzyloxy-pyridine-5-boronic acid, 6-benzyloxy pyridin-3-yl-3-boronic acid, 2-benzyloxypyridin-5-yl boronic acid, 2-benzyloxy pyridine-5-ylboronic acid
17750216
(6-phenylmethoxypyridin-3-yl)boronic acid
OB(O)C1=CN=C(OCC2=CC=CC=C2)C=C1
Melting Point 107°C to 110°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 229.04
Percent Purity 97%
Chemical Name or Material 6-Benzyloxypyridine-3-boronic acid
compliance-icons
Product Identifier
  • 6-Benzyloxypyridine-3-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'