missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-1,2,3,4-tetrahydroisoquinoline, 95%, Thermo Scientific Chemicals

Catalog Number p-7055043
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
226942-29-6
C9H10BrN
212.09
MFCD07374370
URDGCPQHZSDBRG-UHFFFAOYSA-N
isoquinoline, 6-bromo-1,2,3,4-tetrahydro, 6-bromo-1,2,3,4-tetrahydro-isoquinoline, acmc-20ailv, 6-bromo-1,2,3,4-tetra hydro iso quinoline, 6-bromo-1,2,3,4-tetrahydroisoquin, 1,2,3,4-tetrahydro-6-bromoisoquinoline, isoquinoline,6-bromo-1,2,3,4-tetrahydro
15885183
6-bromo-1,2,3,4-tetrahydroisoquinoline
C1CNCC2=C1C=C(C=C2)Br
This item is not returnable. View return policy
226942-29-6
C9H10BrN
212.09
MFCD07374370
URDGCPQHZSDBRG-UHFFFAOYSA-N
isoquinoline, 6-bromo-1,2,3,4-tetrahydro, 6-bromo-1,2,3,4-tetrahydro-isoquinoline, acmc-20ailv, 6-bromo-1,2,3,4-tetra hydro iso quinoline, 6-bromo-1,2,3,4-tetrahydroisoquin, 1,2,3,4-tetrahydro-6-bromoisoquinoline, isoquinoline,6-bromo-1,2,3,4-tetrahydro
15885183
6-bromo-1,2,3,4-tetrahydroisoquinoline
C1CNCC2=C1C=C(C=C2)Br

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 212.09
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 6-Bromo-1,2,3,4-tetrahydroisoquinoline

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'