Learn More
6-Bromo-2,3-dihydrobenzo[b][1,4]dioxine, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 52287-51-1 |
| Molecular Formula | C8H7BrO2 |
| MDL Number | MFCD00040750 |
| Quantity | 100 g |
| SMILES | BrC1=CC=C2OCCOC2=C1 |
| Molecular Weight (g/mol) | 215.04 |
| Percent Purity | 97% |
| Chemical Name or Material | 6-Bromo-2,3-dihydrobenzo[b][1,4]dioxine |