missing translation for 'onlineSavingsMsg'
Learn More

6-Bromo-2,4-dimethylpyridine-3-boronic acid, 95%

Product Code. p-7029937
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15407695 250 mg 250mg
11317329 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15407695 Supplier Thermo Scientific Alfa Aesar Supplier No. H27460.MD
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1072944-23-0 | C7H9BBrNO2 | 229.868 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1072944-23-0
Molecular Formula C7H9BBrNO2
Molecular Weight (g/mol) 229.868
MDL Number MFCD09037491
InChI Key GXXITYQLWOMIEM-UHFFFAOYSA-N
Synonym 6-bromo-2,4-dimethylpyridine-3-boronic acid, 6-bromo-2,4-dimethylpyridin-3-yl boronic acid, acmc-2098oh, 6-bromo-2,4-dimethylpyridin-3-yl boronicacid
PubChem CID 44755186
IUPAC Name (6-bromo-2,4-dimethylpyridin-3-yl)boronic acid
SMILES B(C1=C(N=C(C=C1C)Br)C)(O)O

Specifications

Melting Point ∽145°C (decomposition)
Quantity 250 mg
Formula Weight 229.87
Percent Purity 95%
Chemical Name or Material 6-Bromo-2,4-dimethylpyridine-3-boronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.