Learn More
6-Bromo-2-naphthol, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 6-Bromo-2-naphthol |
| CAS | 15231-91-1 |
| Molecular Formula | C10H7BrO |
| Quantity | 25 g |
| Synonym | 2-Bromo-6-hydroxynaphthalene, 2-Bromo-6-naphthol, 2-Hydroxy-6-bromonaphthalene, 6-Bromo-2-hydroxynaphthalene, 6-Bromo-2-naphthalenol, 6-Bromo-2-naphthalinol, 6-Bromo-2-naphthol, 6-Bromo-beta-naphthol, 6-Bromonaphthol, 6-Hydroxy-2-bromonaphthalene, NSC 17563 |
| InChI Formula | InChI=1S/C10H7BrO/c11-9-3-1-8-6-10(12)4-2-7(8)5-9/h1-6,12H |
| SMILES | Oc1ccc2cc(Br)ccc2c1 |
| IUPAC Name | 6-bromonaphthalen-2-ol |
| Molecular Weight (g/mol) | 223.07 |
| Formula Weight | 221.968 |