Learn More
6-Bromopurine, TRC
Product Code. 30343297
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30343297 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30343297 Supplier Toronto Research Chemicals Supplier No. TRCB6883055G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6-Bromopurine
CAS 767-69-1
Molecular Formula C5H3BrN4
Quantity 5 g
Synonym 6-Bromo-1H-purine, 6-Bromo-9H-purine, 6-Bromopurine, NSC 45150
InChI Formula InChI=1S/C5H3BrN4/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H,7,8,9,10)
SMILES Brc1ncnc2nc[nH]c12
IUPAC Name 6-bromo-7H-purine
Molecular Weight (g/mol) 199.01
Formula Weight 197.9541