Learn More
6-Chloro-2,4-dimethylnicotinonitrile, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Chloro-2,4-dimethylnicotinonitrile |
| CAS | 101251-72-3 |
| Molecular Formula | C8H7N2Cl |
| Quantity | 10 mg |
| InChI Formula | InChI=1S/C8H7ClN2/c1-5-3-8(9)11-6(2)7(5)4-10/h3H,1-2H3 |
| SMILES | CC1=CC(=NC(=C1C#N)C)Cl |
| IUPAC Name | 6-chloro-2,4-dimethylpyridine-3-carbonitrile |
| Molecular Weight (g/mol) | 166.6 |
| Formula Weight | 166.0298 |