Learn More
6-Chloro-2-fluoro-3-methylbenzonitrile, 97%, Thermo Scientific™
Product Code. 11435990
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11435990 1 g 1g
11445990 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11435990 Supplier Thermo Scientific Alfa Aesar Supplier No. H26558.03
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 886502-19-8
Molecular Formula C8H5ClFN
Molecular Weight (g/mol) 169.583
MDL Number MFCD06660186
InChI Key ZZIYYBYHLBGXCD-UHFFFAOYSA-N
Synonym benzonitrile,6-chloro-2-fluoro-3-methyl, 6-chloro-2-fluoro-3-methylbenzenecarbonitrile
PubChem CID 17750749
IUPAC Name 6-chloro-2-fluoro-3-methylbenzonitrile
SMILES CC1=C(C(=C(C=C1)Cl)C#N)F
Specifications
Melting Point 56°C to 58°C
Quantity 1 g
UN Number UN3439
Formula Weight 169.59
Percent Purity 97%
Chemical Name or Material 6-Chloro-2-fluoro-3-methylbenzonitrile