missing translation for 'onlineSavingsMsg'
Learn More

6-chloro-3-(hydroxymethyl)pyridine, 98%, Thermo Scientific™

Catalog Number p-4258900
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
21543-49-7
C6H6ClNO
143.57
GOXYBEXWMJZLJB-UHFFFAOYSA-N
2-chloro-5-hydroxymethylpyridine, 6-chloropyridin-3-yl methanol, 2-chloro-5-hydroxymethyl pyridine, 2-chloro-5-pyridinemethanol, 6-chloropyridine-3-methanol, 6-chloro-3-pyridinemethanol, 6-chloro-3-pyridinyl methanol, 3-pyridinemethanol, 6-chloro, 6-chloro-3-pyridyl methanol, 6-chloro-3-pyridyl methan-1-ol
177161
(6-chloropyridin-3-yl)methanol
C1=CC(=NC=C1CO)Cl
This item is not returnable. View return policy
21543-49-7
C6H6ClNO
143.57
GOXYBEXWMJZLJB-UHFFFAOYSA-N
2-chloro-5-hydroxymethylpyridine, 6-chloropyridin-3-yl methanol, 2-chloro-5-hydroxymethyl pyridine, 2-chloro-5-pyridinemethanol, 6-chloropyridine-3-methanol, 6-chloro-3-pyridinemethanol, 6-chloro-3-pyridinyl methanol, 3-pyridinemethanol, 6-chloro, 6-chloro-3-pyridyl methanol, 6-chloro-3-pyridyl methan-1-ol
177161
(6-chloropyridin-3-yl)methanol
C1=CC(=NC=C1CO)Cl
CAS Min % 97.5
CAS Max % 100.0
Melting Point 46.7°C to 48.5°C
Color White to Brown
Density 1.2800g/mL
Boiling Point 190.0°C
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Specific Gravity 1.28
Formula Weight 143.57
Percent Purity 98%
Physical Form Powder or Low Melting Solid
Chemical Name or Material 6-chloro-3-(hydroxymethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-5-hydroxymethylpyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'