missing translation for 'onlineSavingsMsg'
Learn More

6’-Chloro-norlaudanosoline Hydrobromide, TRC

Product Code. 30352586
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30352586 25 mg -
30359269 10 mg 10mg
30351417 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30352586 Supplier Toronto Research Chemicals Supplier No. TRCC37475025MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C16 H16 Cl N O4 . HBr
Molecular Weight (g/mol) 402.668
Synonym 1-(2-Chloro-4,5-dihydroxybenzyl)-1,2,3,4-tetrahydroisoquinoline-6,7-diol Hydrobromide
IUPAC Name 1-[(2-chloro-4,5-dihydroxyphenyl)methyl]-1,2,3,4-tetrahydroisoquinoline-6,7-diol;hydrobromide
SMILES Br.Oc1cc(Cl)c(CC2NCCc3cc(O)c(O)cc23)cc1O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C16H16ClNO4.BrH/c17-11-7-16(22)14(20)5-9(11)3-12-10-6-15(21)13(19)4-8(10)1-2-18-12;/h4-7,12,18-22H,1-3H2;1H
Formula Weight 401.003
Chemical Name or Material 6’-Chloro-norlaudanosoline Hydrobromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.