missing translation for 'onlineSavingsMsg'
Learn More

6-chloroacetyl-2-benzoxazolinone, 97%, Thermo Scientific™

Catalog Number 11986681
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
54903-10-5
C9H6ClNO3
211.6
KOOWFDRPKXUCRF-UHFFFAOYSA-N
6-chloroacetyl-2-benzoxazolinone, jpb 24, 2 3h-benzoxazolone, 6-chloroacetyl, 6-2-chloroacetyl benzo d oxazol-2 3h-one, 6-chloroacetyl-2 3h-benzoxazolone, 6-2-chloroacetyl-2,3-dihydro-1,3-benzoxazol-2-one, 6-2-chloroacetyl-3-hydrobenzoxazol-2-one, 6-2-chloroacetyl-3h-1,3-benzoxazol-2-one, acmc-20alz8, 6-chloroacetyl benzoxazole-2 3h-one
1988981
6-(2-chloroacetyl)-3H-1,3-benzoxazol-2-one
C1=CC2=C(C=C1C(=O)CCl)OC(=O)N2
This item is not returnable. View return policy
54903-10-5
C9H6ClNO3
211.6
KOOWFDRPKXUCRF-UHFFFAOYSA-N
6-chloroacetyl-2-benzoxazolinone, jpb 24, 2 3h-benzoxazolone, 6-chloroacetyl, 6-2-chloroacetyl benzo d oxazol-2 3h-one, 6-chloroacetyl-2 3h-benzoxazolone, 6-2-chloroacetyl-2,3-dihydro-1,3-benzoxazol-2-one, 6-2-chloroacetyl-3-hydrobenzoxazol-2-one, 6-2-chloroacetyl-3h-1,3-benzoxazol-2-one, acmc-20alz8, 6-chloroacetyl benzoxazole-2 3h-one
1988981
6-(2-chloroacetyl)-3H-1,3-benzoxazol-2-one
C1=CC2=C(C=C1C(=O)CCl)OC(=O)N2
CAS Min % 96.0
CAS Max % 100.0
Melting Point 263.7°C to 269.6°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Formula Weight 211.6
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 6-chloroacetyl-2-benzoxazolinone
Show More Show Less
compliance-icons
Product Identifier
  • 6-Chloroacetyl-2-benzoxazolinone, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'