Learn More
6-Chlorohexanoic acid, 96%, BLD Pharm™
Specifications
Specifications
| CAS | 4224-62-8 |
| Molecular Formula | C6H11ClO2 |
| MDL Number | MFCD00061103 |
| Quantity | 1 g |
| SMILES | OC(=O)CCCCCCl |
| Molecular Weight (g/mol) | 150.60 |
| Percent Purity | 96% |
| Chemical Name or Material | 6-Chlorohexanoic acid |