missing translation for 'onlineSavingsMsg'
Learn More

6-cyclohexylhexanoic Acid, TRC

Product Code. 30366938
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30366938 50 mg -
30353803 100 mg 100mg
30365552 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30366938 Supplier Toronto Research Chemicals Supplier No. TRCC99456550MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4354-56-7
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.3
IUPAC Name 6-cyclohexylhexanoic acid
SMILES OC(=O)CCCCCC1CCCCC1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C12H22O2/c13-12(14)10-6-2-5-9-11-7-3-1-4-8-11/h11H,1-10H2,(H,13,14)
Formula Weight 198.162
Chemical Name or Material 6-cyclohexylhexanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.