missing translation for 'onlineSavingsMsg'
Learn More

6-Dehydrocortisol 21-Hydrogen Succinate, TRC

Product Code. 30351142
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
250mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30351142 250 mg 250mg
30366422 500 mg 500mg
30359117 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30351142 Supplier Toronto Research Chemicals Supplier No. TRCD230644250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Dehydrocortisol 21-Hydrogen Succinate
Quantity 250 mg
Shipping Condition Room Temperature
Molecular Formula C25 H32 O8
Formula Weight 460.21
Molecular Weight (g/mol) 460.517
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.