missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 6-FAM SE

Product Code. 16496996
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16496996

Brand: MedChemExpress HY15939

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress 6-FAM SE is another isomer of carboxyfluorescein.

Specifications

Quantity 100 mg
Molecular Weight (g/mol) 473.39 g/mol
Solubility DMSO : 50 mg/mL (105.62 mM; Need ultrasonic)
Content And Storage -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
CAS 92557-81-8
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.