missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Hydroxy Nicotinic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | Nicotinic acid-6-hydroxy |
| CAS | 5006-66-6 |
| Molecular Formula | C6 H5 N O3 |
| Quantity | 1 g |
| Synonym | 3-Pyridinecarboxylic acid, 1,6-dihydro-6-oxo-, Nicotinic acid, 1,6-dihydro-6-oxo- (8CI), Nicotinic acid, 6-hydroxy- (6CI,7CI), 1,6-Dihydro-6-oxo-3-pyridinecarboxylic acid, 2-Hydroxy-5-carboxypyridine, 2-Hydroxypyridine-5-carboxylic acid, 2-Pyridone-5-carboxylic acid, 5-Carboxy-2-pyridone, 6-Hydroxyniacin, 6-Hydroxynicotinic acid, 6-Hydroxypyridine-3-carboxylic acid, 6-Oxo-1,6-dihydro-pyridine-3-carboxylic acid, NSC 35054, NSC 53377, NSC 8620 |
| InChI Formula | InChI=1S/C6H5NO3/c8-5-2-1-4(3-7-5)6(9)10/h1-3H,(H,7,8)(H,9,10) |
| SMILES | OC(=O)c1ccc(O)nc1 |
| IUPAC Name | 6-hydroxypyridine-3-carboxylic acid |
| Molecular Weight (g/mol) | 139.11 |
| Formula Weight | 139.0269 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?