missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Hydroxynicotinic Acid, Mikromol™
Description
6-Hydroxynicotinic Acid
Specifications
Specifications
| Chemical Name or Material | Nicotinic acid-6-hydroxy |
| CAS | 5006-66-6 |
| Analyte or Component Names | Nicotinic acid-6-hydroxy |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H5 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 3-Pyridinecarboxylic acid, 1,6-dihydro-6-oxo-, Nicotinic acid, 1,6-dihydro-6-oxo- (8CI), Nicotinic acid, 6-hydroxy- (6CI,7CI), 1,6-Dihydro-6-oxo-3-pyridinecarboxylic acid, 2-Hydroxy-5-carboxypyridine, 2-Hydroxypyridine-5-carboxylic acid, 2-Pyridone-5-carboxylic acid, 5-Carboxy-2-pyridone, 6-Hydroxyniacin, 6-Hydroxynicotinic acid, 6-Hydroxypyridine-3-carboxylic acid, 6-Oxo-1,6-dihydro-pyridine-3-carboxylic acid, NSC 35054, NSC 53377, NSC 8620 |
| InChI Formula | InChI=1S/C6H5NO3/c8-5-2-1-4(3-7-5)6(9)10/h1-3H,(H,7,8)(H,9,10) |
| SMILES | OC(=O)c1ccc(O)nc1 |
| IUPAC Name | 6-hydroxypyridine-3-carboxylic acid |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?