missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxynicotinic Acid, Mikromol™

Product Code. 30461537
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30461537 4 x 25 mg -
30050230 25 mg 25mg
17901596 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30461537 Supplier Mikromol Supplier No. MM0553.1500254AMP

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

6-Hydroxynicotinic Acid, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicotinic acid-6-hydroxy
CAS 5006-66-6
Analyte or Component Names Nicotinic acid-6-hydroxy
Shipping Condition Room Temperature
Molecular Formula C6 H5 N O3
Certifications/Compliance ISO 17025
Synonym 3-Pyridinecarboxylic acid, 1,6-dihydro-6-oxo-, Nicotinic acid, 1,6-dihydro-6-oxo- (8CI), Nicotinic acid, 6-hydroxy- (6CI,7CI), 1,6-Dihydro-6-oxo-3-pyridinecarboxylic acid, 2-Hydroxy-5-carboxypyridine, 2-Hydroxypyridine-5-carboxylic acid, 2-Pyridone-5-carboxylic acid, 5-Carboxy-2-pyridone, 6-Hydroxyniacin, 6-Hydroxynicotinic acid, 6-Hydroxypyridine-3-carboxylic acid, 6-Oxo-1,6-dihydro-pyridine-3-carboxylic acid, NSC 35054, NSC 53377, NSC 8620
InChI Formula InChI=1S/C6H5NO3/c8-5-2-1-4(3-7-5)6(9)10/h1-3H,(H,7,8)(H,9,10)
Solution Type Neat
SMILES OC(=O)c1ccc(O)nc1
IUPAC Name 6-hydroxypyridine-3-carboxylic acid
Molecular Weight (g/mol) 139.11
Quantity 4 x 25 mg
Formula Weight 139.0269 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.