missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Mercaptopurine, MedChemExpress
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
Description
6-Mercaptopurine is a purine analogue which acts as an antagonist of the endogenous purines and has been widely used as antileukemic agent and immunosuppressive drug.
Specifications
Specifications
| Chemical Name or Material | 6-Mercaptopurine |
| Physical Form | Solid |
| CAS | 50-44-2 |
| Color | Yellow |
| Quantity | 500 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C5H4N4S |
| For Use With (Application) | Cancer-programmed cell death |
| Synonym | Mercaptopurine 6-MP |
| Solubility Information | DMSO : 35.71 mg/mL (234.66 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction