Learn More
6-Methoxy-1-indanone, 98%, Thermo Scientific Chemicals
Catalog Number p-4510581
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
13623-25-1
C10H10O2
162.19
MFCD00021232
UJGDLLGKMWVCPT-UHFFFAOYSA-N
6-methoxy-1-indanone, 6-methoxy-1h-indanone, 6-methoxy-indan-1-one, 6-methoxy-2,3-dihydro-1h-inden-1-one, 6-methoxyindan-1-one, 2,3-dihydro-6-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-6-methoxy, 6-methoxyindanone, 6-methoxy indanone, 6-methoxy-1-indanon
334036
6-methoxy-2,3-dihydroinden-1-one
COC1=CC=C2CCC(=O)C2=C1
13623-25-1
C10H10O2
162.19
MFCD00021232
UJGDLLGKMWVCPT-UHFFFAOYSA-N
6-methoxy-1-indanone, 6-methoxy-1h-indanone, 6-methoxy-indan-1-one, 6-methoxy-2,3-dihydro-1h-inden-1-one, 6-methoxyindan-1-one, 2,3-dihydro-6-methoxyinden-1-one, 1h-inden-1-one, 2,3-dihydro-6-methoxy, 6-methoxyindanone, 6-methoxy indanone, 6-methoxy-1-indanon
334036
6-methoxy-2,3-dihydroinden-1-one
COC1=CC=C2CCC(=O)C2=C1
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Melting Point 107.0°C to 111.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethyl acetate,methylene dichloride
Formula Weight 162.19
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 6-Methoxy-1-indanone
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methoxy-1-indanone
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.