missing translation for 'onlineSavingsMsg'
Learn More

6-Methoxy-2-naphthonitrile, 98%, Thermo Scientific™

Catalog Number 11975611
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
67886-70-8
C12H9NO
183.21
SWEFYGCKCMJSPD-UHFFFAOYSA-N
6-methoxy-2-naphthonitrile, 2-cyano-6-methoxynaphthalene, 6-methoxy-2-cyanonaphthalene, 6-methoxy-2-naphthalenecarbonitrile, 6-methoxy-naphthalene-2-carbonitrile, 2-naphthalenecarbonitrile,6-methoxy, acmc-20anxg, 2-cyan-6-methoxy-naphthalin, 2-naphthonitrile, 6-methoxy
349173
6-methoxynaphthalene-2-carbonitrile
COC1=CC2=C(C=C1)C=C(C=C2)C#N
This item is not returnable. View return policy
67886-70-8
C12H9NO
183.21
SWEFYGCKCMJSPD-UHFFFAOYSA-N
6-methoxy-2-naphthonitrile, 2-cyano-6-methoxynaphthalene, 6-methoxy-2-cyanonaphthalene, 6-methoxy-2-naphthalenecarbonitrile, 6-methoxy-naphthalene-2-carbonitrile, 2-naphthalenecarbonitrile,6-methoxy, acmc-20anxg, 2-cyan-6-methoxy-naphthalin, 2-naphthonitrile, 6-methoxy
349173
6-methoxynaphthalene-2-carbonitrile
COC1=CC2=C(C=C1)C=C(C=C2)C#N
CAS Min % 97.5
CAS Max % 100.0
Melting Point 106.0°C to 107.0°C
Color White to Yellow
Boiling Point 205.0°C to 208.0°C (14.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Linear Formula CH3OC10H6CN
Quantity 5 g
Beilstein 10,III,1099
Formula Weight 183.21
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 6-Methoxy-2-naphthonitrile
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methoxy-2-naphthonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'