missing translation for 'onlineSavingsMsg'
Learn More
Learn More
6-Methoxy-N-(3-sulfopropyl)quinolinium, TRC
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Specifications
Specifications
| Chemical Name or Material | 6-Methoxy-N-(3-sulfopropyl)quinolinium |
| CAS | 83907-40-8 |
| Molecular Formula | C13 H15 N O4 S |
| Quantity | 100 mg |
| Synonym | Quinolinium','6-methoxy-1-(3-sulfopropyl)-','inner salt, SPQ |
| InChI Formula | InChI=1S/C13H15NO4S/c1-18-12-5-6-13-11(10-12)4-2-7-14(13)8-3-9-19(15,16)17/h2,4-7,10H,3,8-9H2,1H3 |
| SMILES | COc1ccc2c(ccc[n+]2CCCS(=O)(=O)[O-])c1 |
| IUPAC Name | 3-(6-methoxyquinolin-1-ium-1-yl)propane-1-sulfonate |
| Molecular Weight (g/mol) | 281.33 |
| Formula Weight | 281.0722 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction