Learn More
6-Monoamino-Beta-cyclodextrin, TRC
Product Code. 30470629
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30470629 10 mg 10mg
30465707 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30470629 Supplier Toronto Research Chemicals Supplier No. TRCM52481310MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 29390-67-8
Molecular Formula C42H71NO34
Molecular Weight (g/mol) 1134
Synonym 6A-Amino-6A-deoxy-beta-cyclodextrin, 6-Amino-6-deoxy-beta-cyclodextrin, 6-Amino-6-deoxycyclomaltoheptaose, 6-Deoxy-6-amino-beta-cyclodextrin, 6-Monoamino-6-monodeoxy-beta-cyclodextrin, 6-Monodeoxy-6-monoamino-beta-cyclodextrin, Mono(6-amino-6-deoxy)-beta-cyclodextrin, Mono(6-deoxy-6-amino)-beta-cyclodextrin
SMILES NC[C@H]1O[C@@H]2O[C@H]3[C@H](O)[C@@H](O)[C@H](O[C@@H]3CO)O[C@H]4[C@H](O)[C@@H](O)[C@H](O[C@@H]4CO)O[C@H]5[C@H](O)[C@@H](O)[C@H](O[C@@H]5CO)O[C@H]6[C@H](O)[C@@H](O)[C@H](O[C@@H]6CO)O[C@H]7[C@H](O)[C@@H](O)[C@H](O[C@@H]7CO)O[C@H]8[C@H](O)[C@@H](O)[C@H](O[C@@H]8CO)O[C@H]1[C@H](O)[C@H]2O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C42H71NO34/c43-1-8-29-15(50)22(57)36(64-8)72-30-9(2-44)66-38(24(59)17(30)52)74-32-11(4-46)68-40(26(61)19(32)54)76-34-13(6-48)70-42(28(63)21(34)56)77-35-14(7-49)69-41(27(62)20(35)55)75-33-12(5-47)67-39(25(60)18(33)53)73-31-10(3-45)65-37(71-29)23(58)16(31)51/h8-42,44-63H,1-7,43H2/t8-,9-,10-,11-,12-,13-,14-,15-,16-,17-,18-,19-,20-,21-,22-,23-,24-,25-,26-,27-,28-,29-,30-,31-,32-,33-,34-,35-,36-,37-,38-,39-,40-,41-,42-/m1/s1
Formula Weight 1133.3857
Chemical Name or Material 6-Monoamino-beta-cyclodextrin