missing translation for 'onlineSavingsMsg'
Learn More

6 Phosphofructo 2 Kinase Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP1-87478

 View more versions of this product

Product Code. 18447041

  • € 725.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

6 Phosphofructo 2 Kinase Polyclonal antibody specifically detects 6 Phosphofructo 2 Kinase in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 Phosphofructo 2 Kinase
Polyclonal
Western Blot 0.04-0.4 ug/mL, Immunocytochemistry/ Immunofluorescence 0.25-2 ug/mL
Q16875
Rabbit
Immunogen affinity purified
RUO
Primary
Human
Purified
Western Blot, Immunocytochemistry
Unconjugated
PBS (pH 7.2) and 40% Glycerol
6PF-2-K/Fru-2,6-P2ase 3,6-bisphosphatase, 6-phosphofructo-2-kinase/ fructose-2,6-bisphosphatase, 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 3,6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase, FLJ37326, inducible 6-phosphofructo-2-kinase/fructose-2,6-bisphosphatase, IPFK2, Renal carcinoma antigen NY-REN-56
This antibody was developed against Recombinant Protein corresponding to amino acids:EEAMKVRKQCALAALRDVKSYLAKEGGQIAVFDATNTTRERRHMILHFAKENDFKAFFIESVCDDPTVVASNIMEVKISSPDYKDCNSAEAMDDFMKRISCYEASYQPLDPDKCDRDLSLIKVIDVGRRFLVNRVQD
0.1 mL
Endocrinology
5209
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.