Learn More
MedChemExpress 6-Thio-2'-Deoxyguanosine
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Specifications
Specifications
| Molecular Weight (g/mol) | 283.31 g/mol |
| Product Type | Reference compound |
| Content And Storage | 4°C, protect from light∣In solvent : -80°C, 6 months∣-20°C, 1 month (protect from light) |
| Shipping Condition | Room temperature |
| Purity | 97.0% |
| Quantity | 5 mg |
| Solubility Information | DMSO : 50 mg/mL (176.49 mM∣Need ultrasonic) |
| For Use With (Application) | Cancer-programmed cell death |
| CAS | 789-61-7 |
| Synonym | 6-thio-dG∣ β-TGdR |