missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ 6-Tube Format Autotube for MPure-32

Product Code. 17722528 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1536/Pk.
Unit Size:
Pack of 1536
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17722528 1536/Pk. Pack of 1536
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17722528 Supplier MP Biomedicals™ Supplier No. 117032149
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

6-Tube Format Autotube to use with 16-Base on MPure-32 aNAP system. Ideal for 1 to 15 runs. Each 6-well rack is designed to receive the sample and reagents for one run.

  • To use with 16-Base on MPure-32 aNAP system.
  • Ideal for 1 to 15 runs.
  • Each 6-well rack is designed to receive the sample and reagents for one run.
TRUSTED_SUSTAINABILITY

Specifications

Sample Type Tissue, Cell Culture, Bacteria
Format Tube
For Use With (Equipment) MPure-32
Quantity 1536/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.