Learn More
Poeppelmann™ 600 Series Natural LDPE Tapered Closures
Product Code. 10492545
Change view
Click to view available options
Diameter (Metric) Outside:
189 mm
Unit Size:
Pack of 25
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Outside Unit Size
10492545 189 mm Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10492545 Supplier Poeppelmann™ Supplier No. 60017900000
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Specifications
Color Natural
For Use With (Application) For use as plugs in bores or internal thread, or as caps for external thread
Length (Metric) 23 mm
Quantity 25 Pack
Diameter (Metric) Outside 189 mm
Material LDPE
Type Tapered Closure