missing translation for 'onlineSavingsMsg'
Learn More

3M™ 6000 Series Half Face Mask Respirator

Product Code. 12642555
Click to view available options
Size:
Small
Medium
For Use With (Application):
Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
Product Type:
Half-Mask Respirator
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12642555

Brand: 3M™ 7000146847

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Requires

Facepiece requires compatible cartridges and/or filters. (Sold separately.)

Half-face mask respirators 6000 Series are economical, low-maintenance, and extremely lightweight. Probed reusable respirator assembly 6000Q series are used for quantitative fit purposes only.

Combines features of both reusable and disposable respirators to provide effective protection. 3M™ Half Face Mask Respirator 6000 Series are economical, low-maintenance, simple-to-handle, and extremely lightweight.

  • Reusable half masks are economical, low-maintenance, simple-to-handle, and extremely lightweight
  • Used with 3M™ Supplied Air Systems
  • Bayonet Connection System allows connection to a broad range of twin lightweight filters to protect against gases, vapors, and particulates
  • Twin filter design provides lower breathing resistance
  • Probed reusable respirator assembly 6000 Series used for quantitative fit testing (17-987-3A & -3B)

Order Info

Probed models for fit test only—not for use as a respirator. Cartridge/filter combinations sold separately.

TRUSTED_SUSTAINABILITY

Specifications

Certifications (EN) EN 140:1998
Certifications (NIOSH) NIOSH 42 CFR Part 84
Connection Type Bayonet
Color Gray
Material Thermoplastic Elastomer
Product Type Half-Mask Respirator
Size Medium
For Use With (Application) Assembly, cleaning, demolition, facility maintenance, grinding, machine operation, painting, remediation, sanding, welding
For Use With (Equipment) Cartridge 6000 Series, Filter 2000, 2200, 7000 or 5000 Series, or Dual Airline System
Includes Reclosable storage bag
Model 6200
Suspension Type 4-point
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.