missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ 60S ribosomal protein L23 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-29994
This item is not returnable.
View return policy
Description
60S ribosomal protein L23 Polyclonal antibody specifically detects 60S ribosomal protein L23 in Human,Mouse samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| 60S ribosomal protein L23 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 9349 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| MGC111167, MGC117346, MGC72008,60S ribosomal protein L17,60S ribosomal protein L23, ribosomal protein L17, ribosomal protein L23, rpL17 | |
| Between 90 and 140 | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction