Learn More
6beta-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate (6beta-Hydroxymedroxyprogesterone Acetate), Mikromol™
Product Code. 30052046
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Product Code. 30052046
Brand: Mikromol MM0532.010025
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
6beta-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate (6beta-Hydroxymedroxyprogesterone Acetate)
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 6beta-Hydroxymedroxyprogesterone Acetate
CAS 984-47-4
Analyte or Component Names 6beta-Hydroxymedroxyprogesterone Acetate
Shipping Condition Room Temperature
Molecular Formula C24 H34 O5
Certifications/Compliance ISO 17025
Synonym 6beta-Hydroxy-6-methyl-3,20-dioxopregn-4-en-17-yl Acetate, Medroxyprogesterone Acetate Imp. A (EP), 6beta-Hydroxymedroxyprogesterone Acetate, Pregn-4-ene-3,20-dione, 17-(acetyloxy)-6-hydroxy-6-methyl-, (6beta)-, Pregn-4-ene-3,20-dione, 6beta,17-dihydroxy-6-methyl-, 17-acetate (7CI,8CI), (6beta)-17-(Acetyloxy)-6-hydroxy-6-methylpregn-4-ene-3,20-dione, Medroxyprogesterone Acetate Impurity A
InChI Formula InChI=1S/C24H34O5/c1-14(25)24(29-15(2)26)11-8-19-17-13-23(5,28)20-12-16(27)6-9-21(20,3)18(17)7-10-22(19,24)4/h12,17-19,28H,6-11,13H2,1-5H3/t17-,18+,19+,21-,22+,23-,24+/m1/s1
SMILES CC(=O)O[C@@]1(CC[C@H]2[C@@H]3C[C@@](C)(O)C4=CC(=O)CC[C@]4(C)[C@H]3CC[C@]12C)C(=O)C
IUPAC Name [(6R,8R,9S,10R,13S,14S,17R)-17-acetyl-6-hydroxy-6,10,13-trimethyl-3-oxo-1,2,7,8,9,11,12,14,15,16-decahydrocyclopenta[a]phenanthren-17-yl] acetate
Molecular Weight (g/mol) 402.52
Quantity 25 mg
Formula Weight 402.2406 g/mol