missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(6R,12aS)-6-(1,3-Benzodioxol-5-yl)-2-methyl-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
(6R,12aS)-6-(1,3-Benzodioxol-5-yl)-2-methyl-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione
Specifications
Specifications
| Chemical Name or Material | (6R,12aS)-6-(1,3-Benzodioxol-5-yl)-2-methyl-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione |
| CAS | 171596-27-3 |
| Analyte or Component Names | (6R,12aS)-6-(1,3-Benzodioxol-5-yl)-2-methyl-2,3,6,7,12,12a-hexahydropyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione |
| Shipping Condition | Room Temperature |
| Molecular Formula | C22 H19 N3 O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | (6R,12aS)-6-(1,3-Benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-methylpyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, Pyrazino[1',2':1,6]pyrido[3,4-b]indole-1,4-dione, 6-(1,3-benzodioxol-5-yl)-2,3,6,7,12,12a-hexahydro-2-methyl-, (6R-cis)-, 12-epi-Tadalafil, Tadalafil Imp. A (EP), 6R,12S-Tadalafil, Tadalafil Impurity A |
| InChI Formula | InChI=1S/C22H19N3O4/c1-24-10-19(26)25-16(22(24)27)9-14-13-4-2-3-5-15(13)23-20(14)21(25)12-6-7-17-18(8-12)29-11-28-17/h2-8,16,21,23H,9-11H2,1H3/t16-,21+/m0/s1 |
| SMILES | CN1CC(=O)N2[C@@H](Cc3c([nH]c4ccccc34)[C@H]2c5ccc6OCOc6c5)C1=O |
| IUPAC Name | (2R,8S)-2-(1,3-benzodioxol-5-yl)-6-methyl-3,6,17-triazatetracyclo[8.7.0.0^{3,8}.0^{11,16}]heptadeca-1(10),11,13,15-tetraene-4,7-dione |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction